Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-N-Fmoc-L-phenylalanine, 97%

Chemical Identifiers

Chemical Name or Material 2-Chloro-N-Fmoc-L-phenylalanine
CAS 198560-41-7
Formula Weight 421.88
MDL Number MFCD00797578
Beilstein 10203122
Synonym Fmoc-Phe(2-Cl)-OH; (S)-3-(2-Chlorophenyl)-2-(Fmoc-amino)propionic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6625814

 
alfa aesar™
H6625814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6625806

 
alfa aesar™
H6625806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 421.88
CAS 198560-41-7
Chemical Name or Material 2-Chloro-N-Fmoc-L-phenylalanine
MDL Number MFCD00797578
Synonym Fmoc-Phe(2-Cl)-OH; (S)-3-(2-Chlorophenyl)-2-(Fmoc-amino)propionic acid
Assay 97%
Physical Form Powder
Storage Note 1 2° to 8°C
Color White
Quantity 25g
Beilstein 10203122
Safety and Handling

2° to 8°C