Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-N-cyclohexylnicotinamide, 99%

$33.91 - $65.69

Chemical Identifiers

Chemical Name or Material 2-Chloro-N-cyclohexylnicotinamide
CAS 57841-70-0
Molecular Formula C12H15ClN2O
Formula Weight 238.72
MDL Number MFCD01344059
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5053303

  SDS
alfa aesar™
H5053303
1g Each for $33.91

AAH5053306

  SDS
alfa aesar™
H5053306
5g Each for $65.69
Description & Specifications

Specifications

Formula Weight 238.72
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Molecular Formula C12H15ClN2O
Quantity 1g
Chemical Name or Material 2-Chloro-N-cyclohexylnicotinamide
Assay 99%
CAS 57841-70-0
MDL Number MFCD01344059
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.