missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-N-(3-methylphenyl)-4-nitrobenzamide, 97%

Chemical Identifiers

CAS 329940-70-7
Molecular Formula C14H11ClN2O3
Molecular Weight (g/mol) 290.703
MDL Number MFCD00784494
InChI Key VIQSGUZRECTJEM-UHFFFAOYSA-N
Synonym 2-chloro-4-nitro-n-3-methylphenyl benzamide, 2-chloro-4-nitrophenyl-n-3-methylphenyl carboxamide, 2-chloro-n-3-methylphenyl-4-nitrobenzamide, cambridge id 5344357
PubChem CID 710885
IUPAC Name 2-chloro-N-(3-methylphenyl)-4-nitrobenzamide
SMILES CC1=CC(=CC=C1)NC(=O)C2=C(C=C(C=C2)[N+](=O)[O-])Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH59987MD View Documents Alfa Aesar™
H59987MD
250mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH5998703 View Documents Alfa Aesar™
H5998703
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

329940-70-7
290.703
VIQSGUZRECTJEM-UHFFFAOYSA-N
710885
CC1=CC(=CC=C1)NC(=O)C2=C(C=C(C=C2)[N+](=O)[O-])Cl
C14H11ClN2O3
MFCD00784494
2-chloro-4-nitro-n-3-methylphenyl benzamide, 2-chloro-4-nitrophenyl-n-3-methylphenyl carboxamide, 2-chloro-n-3-methylphenyl-4-nitrobenzamide, cambridge id 5344357
2-chloro-N-(3-methylphenyl)-4-nitrobenzamide

Specifications

250mg
C14H11ClN2O3
2-chloro-4-nitro-n-3-methylphenyl benzamide, 2-chloro-4-nitrophenyl-n-3-methylphenyl carboxamide, 2-chloro-n-3-methylphenyl-4-nitrobenzamide, cambridge id 5344357
CC1=CC(=CC=C1)NC(=O)C2=C(C=C(C=C2)[N+](=O)[O-])Cl
290.703
290.7
2-Chloro-N-(3-methylphenyl)-4-nitrobenzamide
329940-70-7
MFCD00784494
VIQSGUZRECTJEM-UHFFFAOYSA-N
2-chloro-N-(3-methylphenyl)-4-nitrobenzamide
710885
97%
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures