missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-N-(2,5-dimethoxyphenyl)benzamide, 97%

$211.27 - $675.45

Chemical Identifiers

CAS 196938-30-4
Molecular Formula C15H14ClNO3
Molecular Weight (g/mol) 291.731
MDL Number MFCD00095371
InChI Key RLHACPNMKPCNMS-UHFFFAOYSA-N
Synonym 2-chloro-n-2,5-dimethoxyphenyl benzamide, benzamide, n-2,5-dimethoxyphenyl-2-chloro, 2-chloro-2',5'-dimethoxybenzanilide
PubChem CID 307419
IUPAC Name 2-chloro-N-(2,5-dimethoxyphenyl)benzamide
SMILES COC1=CC(=C(C=C1)OC)NC(=O)C2=CC=CC=C2Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH59906MD View Documents Alfa Aesar™
H59906MD
250mg Each for $211.27
 
AAH5990603 View Documents Alfa Aesar™
H5990603
1g Each for $675.45
 
Specifications

Chemical Identifiers

196938-30-4
291.731
RLHACPNMKPCNMS-UHFFFAOYSA-N
307419
COC1=CC(=C(C=C1)OC)NC(=O)C2=CC=CC=C2Cl
C15H14ClNO3
MFCD00095371
2-chloro-n-2,5-dimethoxyphenyl benzamide, benzamide, n-2,5-dimethoxyphenyl-2-chloro, 2-chloro-2',5'-dimethoxybenzanilide
2-chloro-N-(2,5-dimethoxyphenyl)benzamide

Specifications

250mg
C15H14ClNO3
2-chloro-n-2,5-dimethoxyphenyl benzamide, benzamide, n-2,5-dimethoxyphenyl-2-chloro, 2-chloro-2',5'-dimethoxybenzanilide
COC1=CC(=C(C=C1)OC)NC(=O)C2=CC=CC=C2Cl
291.731
291.73
2-Chloro-N-(2,5-dimethoxyphenyl)benzamide
196938-30-4
MFCD00095371
RLHACPNMKPCNMS-UHFFFAOYSA-N
2-chloro-N-(2,5-dimethoxyphenyl)benzamide
307419
97%