Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-N-[2-(2-thienyl)ethyl]-D-phenylglycine methyl ester hydrochloride, 98%

Chemical Identifiers

Chemical Name or Material 2-Chloro-N-[2-(2-thienyl)ethyl]-D-phenylglycine methyl ester hydrochloride
CAS 141109-19-5
Formula Weight 346.27
MDL Number MFCD08460122
Synonym Methyl (S)-(+)-α-[2-(2-thienyl)ethylamino](2-chlorophenyl)acetate hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6193603

 
alfa aesar™
H6193603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6193606

 
alfa aesar™
H6193606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH61936MD

 
alfa aesar™
H61936MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 346.27
Chemical Name or Material 2-Chloro-N-[2-(2-thienyl)ethyl]-D-phenylglycine methyl ester hydrochloride
Physical Form Solid
Assay 98%
MDL Number MFCD08460122
CAS 141109-19-5
Synonym Methyl (S)-(+)-α-[2-(2-thienyl)ethylamino](2-chlorophenyl)acetate hydrochloride
Quantity 1g