Please login to your online account to display your discounted pricing

Alfa Aesar™ (+)-2-Chloro-L-phenylglycine, 95%

$53.93 - $195.88

Chemical Identifiers

Chemical Name or Material (+)-2-Chloro-L-phenylglycine
CAS 141315-50-6
Molecular Formula C8H8ClNO2
Formula Weight 185.61
MDL Number MFCD03426346
Synonym H-Phg(2-Cl)-OH; H-Phg(2-Cl)-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2877603

  SDS
alfa aesar™
H2877603
1g Each for $53.93

AAH2877606

  SDS
alfa aesar™
H2877606
5g Each for $195.88
Description & Specifications

Specifications

Formula Weight 185.61
Molecular Formula C8H8ClNO2
Chemical Name or Material (+)-2-Chloro-L-phenylglycine
CAS 141315-50-6
Optical Rotation Not completely soluble in Chloroform
Melting Point 182° to 187°C
Quantity 1g
MDL Number MFCD03426346
Synonym H-Phg(2-Cl)-OH; H-Phg(2-Cl)-OH
Assay 95%