missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-beta-nitrostyrene, 98%

Manufacturer:  Alfa Aesar™ A1488222

 View more versions of this product

Catalog No. AAA1488222



Specifications

Specifications

2-Chloro-beta-nitrostyrene
100g
C8H6ClNO2
2328095
QHKJTRDWAZGBLR-AATRIKPKSA-N
1-chloro-2-[(E)-2-nitroethenyl]benzene
5369313
98%
46°C to 49°C
3156-34-1
MFCD00024820
1-chloro-2-2-nitrovinyl benzene, 2-chloro-omega-nitrostyrene, 2-chloro-b-nitrostyrene, e-1-chloro-2-2-nitrovinyl benzene, 1-chloro-2-e-2-nitrovinyl benzene, trans-2-chloro-beta-nitrostyrene, 2-chloro-beta-nitrostyrene, styrene, o-chloro-.beta.-nitro, benzene,1-chloro-2-2-nitroethenyl, 1-chloro-2-e-2-nitroethenyl benzene
C1=CC=C(C(=C1)C=C[N+](=O)[O-])Cl
183.591
183.6

Chemical Identifiers

3156-34-1
183.591
QHKJTRDWAZGBLR-AATRIKPKSA-N
5369313
C1=CC=C(C(=C1)C=C[N+](=O)[O-])Cl
C8H6ClNO2
MFCD00024820
1-chloro-2-2-nitrovinyl benzene, 2-chloro-omega-nitrostyrene, 2-chloro-b-nitrostyrene, e-1-chloro-2-2-nitrovinyl benzene, 1-chloro-2-e-2-nitrovinyl benzene, trans-2-chloro-beta-nitrostyrene, 2-chloro-beta-nitrostyrene, styrene, o-chloro-.beta.-nitro, benzene,1-chloro-2-2-nitroethenyl, 1-chloro-2-e-2-nitroethenyl benzene
1-chloro-2-[(E)-2-nitroethenyl]benzene
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents