missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chloro-8-cyclopentyl-5-methyl-8H-pyrido[2,3-d]pyrimidin-7-one, 97%, Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific H5436306

 View more versions of this product

Catalog No. AAH5436306



Description

Description

Applications
2-Chloro-8-cyclopentyl-5-methyl-8H-pyrido[2,3-d]pyrimidin-7-one is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

2-Chloro-8-cyclopentyl-5-methyl-8H-pyrido[2,3-d]pyrimidin-7-one
1013916-37-4
MFCD13181207
2-chloro-8-cyclopentyl-5-methyl-7h,8h-pyrido 2,3-d pyrimidin-7-one
BSKNQSYIDZUXQT-UHFFFAOYSA-N
2-chloro-8-cyclopentyl-5-methylpyrido[2,3-d]pyrimidin-7-one
44248248
97%
5g
C13H14ClN3O
15642863
Slightly soluble in water.
CC1=CC(=O)N(C2=NC(=NC=C12)Cl)C3CCCC3
263.725
263.72

Chemical Identifiers

1013916-37-4
263.725
BSKNQSYIDZUXQT-UHFFFAOYSA-N
44248248
CC1=CC(=O)N(C2=NC(=NC=C12)Cl)C3CCCC3
C13H14ClN3O
MFCD13181207
2-chloro-8-cyclopentyl-5-methyl-7h,8h-pyrido 2,3-d pyrimidin-7-one
2-chloro-8-cyclopentyl-5-methylpyrido[2,3-d]pyrimidin-7-one
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents