missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chloro-8-cyclopentyl-5-methyl-8H-pyrido[2,3-d]pyrimidin-7-one, 97%, Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific H54363MD

 View more versions of this product

Catalog No. AAH54363MD



Description

Description

Applications
2-Chloro-8-cyclopentyl-5-methyl-8H-pyrido[2,3-d]pyrimidin-7-one is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

2-Chloro-8-cyclopentyl-5-methyl-8H-pyrido[2,3-d]pyrimidin-7-one
1013916-37-4
MFCD13181207
2-chloro-8-cyclopentyl-5-methyl-7h,8h-pyrido 2,3-d pyrimidin-7-one
CC1=CC(=O)N(C2=NC(=NC=C12)Cl)C3CCCC3
263.725
263.72
250mg
C13H14ClN3O
15642863
BSKNQSYIDZUXQT-UHFFFAOYSA-N
2-chloro-8-cyclopentyl-5-methylpyrido[2,3-d]pyrimidin-7-one
44248248
97%

Chemical Identifiers

1013916-37-4
263.725
BSKNQSYIDZUXQT-UHFFFAOYSA-N
44248248
CC1=CC(=O)N(C2=NC(=NC=C12)Cl)C3CCCC3
C13H14ClN3O
MFCD13181207
2-chloro-8-cyclopentyl-5-methyl-7h,8h-pyrido 2,3-d pyrimidin-7-one
2-chloro-8-cyclopentyl-5-methylpyrido[2,3-d]pyrimidin-7-one
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents