Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-6-(trifluoromethyl)pyridine, 97%

Chemical Identifiers

Chemical Name or Material 2-Chloro-6-(trifluoromethyl)pyridine
CAS 39890-95-4
Molecular Formula C6H3ClF3N
Formula Weight 181.55
MDL Number MFCD00042226
UN Number UN2811
Beilstein 4308910
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2003403

  SDS
alfa aesar™
L2003403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL2003406

  SDS
alfa aesar™
L2003406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL20034MD

  SDS
alfa aesar™
L20034MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 181.55
Chemical Name or Material 2-Chloro-6-(trifluoromethyl)pyridine
MDL Number MFCD00042226
Beilstein 4308910
Health Hazard 2 GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 97%
Molecular Formula C6H3ClF3N
CAS 39890-95-4
UN Number UN2811
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Flash Point 78°C (172°F)
Boiling Point 60° to 62°C (8mmHg)
Melting Point 28° to 29°C
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.