missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chloro-6-(trifluoromethyl)pyridine, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC247210050

 View more versions of this product

Catalog No. AC247210050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-Chloro-6-(trifluoromethyl)pyridine
60.0°C to 62.0°C (8.0mmHg)
5g
94% min. (GC)
2-chloro-6-trifluoromethyl pyridine, 2-chloro-6-trifluoromethyl-pyridine, 6-chloro-2-trifluoromethyl pyridine, 6-chloro-2-trifluoromethylpyridine, 6-chloro-alpha,alpha,alpha-trifluoro-2-picoline, acmc-209j8p, pubchem2080, pyridine, 2-chloro-6-trifluoromethyl
C1=CC(=NC(=C1)Cl)C(F)(F)F
181.54
181.54
79.4°C
33.0°C to 35.0°C
39890-95-4
C6H3ClF3N
ADVQMCQMDHBTHJ-UHFFFAOYSA-N
2-chloro-6-(trifluoromethyl)pyridine
602334
95%

Chemical Identifiers

39890-95-4
181.54
2-chloro-6-trifluoromethyl pyridine, 2-chloro-6-trifluoromethyl-pyridine, 6-chloro-2-trifluoromethyl pyridine, 6-chloro-2-trifluoromethylpyridine, 6-chloro-alpha,alpha,alpha-trifluoro-2-picoline, acmc-209j8p, pubchem2080, pyridine, 2-chloro-6-trifluoromethyl
2-chloro-6-(trifluoromethyl)pyridine
C6H3ClF3N
ADVQMCQMDHBTHJ-UHFFFAOYSA-N
602334
C1=CC(=NC(=C1)Cl)C(F)(F)F
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.
Causes skin irritation.
Harmful if inhaled.
Causes serious eye irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Cal

SDS Documents