Please login to your online account to display your discounted pricing

2-Chloro-6-(trifluoromethyl)pyridine, 95%, Acros Organics

Chemical Identifiers

Chemical Name or Material 2-Chloro-6-(trifluoromethyl)pyridine
Assay Percent Range 95%
Molecular Formula C6H3ClF3N
Formula Weight 181.54
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC247210050

 
thermo fisher scientific
AC247210050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC247210010

 
thermo fisher scientific
AC247210010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 181.54
Quantity 5g
Chemical Name or Material 2-Chloro-6-(trifluoromethyl)pyridine
Molecular Formula C6H3ClF3N
Assay Percent Range 95%
CAS 39890-95-4