Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-6-(trifluoromethyl)nicotinic acid, 97%

$137.86 - $459.57

Chemical Identifiers

Chemical Name or Material 2-Chloro-6-(trifluoromethyl)nicotinic acid
CAS 280566-45-2
Molecular Formula C7H3ClF3NO2
Formula Weight 225.56
MDL Number MFCD01862658
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2680527

  SDS
alfa aesar™
H2680527
2.5g Each for $459.57

AAH26805ME

  SDS
alfa aesar™
H26805ME
500mg Each for $137.86
Description & Specifications

Specifications

Formula Weight 225.56
Molecular Formula C7H3ClF3NO2
Quantity 2.5g
Melting Point 118° to 120°C
Chemical Name or Material 2-Chloro-6-(trifluoromethyl)nicotinic acid
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 280566-45-2
Assay 97%
Health Hazard 1 Warning
MDL Number MFCD01862658
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.