Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-6-(trifluoromethyl)benzoyl chloride, 97%

Chemical Identifiers

Chemical Name or Material 2-Chloro-6-(trifluoromethyl)benzoyl chloride
CAS 916420-44-5
Molecular Formula C8H3Cl2F3O
Formula Weight 243.01
MDL Number MFCD09025361
UN Number UN3265
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3006203

  SDS
alfa aesar™
H3006203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3006206

  SDS
alfa aesar™
H3006206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 243.01
Chemical Name or Material 2-Chloro-6-(trifluoromethyl)benzoyl chloride
Assay 97%
Molecular Formula C8H3Cl2F3O
MDL Number MFCD09025361
CAS 916420-44-5
UN Number UN3265
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
Quantity 1g

Applications
The use of pentafluoropropionic anhydride, trifluoromethylbenzoyl chloride and pentafluorobenzoyl chloride gave fluoro-chitin derivatives with 40, 10 and 5% substitution, respectively. Fluorination of chitin was achieved by facile homogenous reaction of chitin solution with diethyl amino sulfur trifluoride.

Solubility
Insoluble in water.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, reducind agents, bases, water, moisture.

Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.