missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Chloro-6-(trifluoromethyl)benzoic acid, 97%, Thermo Scientific™
$113.68 - $441.45
Chemical Identifiers
CAS | 2376-00-3 |
---|---|
Molecular Formula | C8H4ClF3O2 |
Molecular Weight (g/mol) | 224.563 |
MDL Number | MFCD06208254 |
InChI Key | OHWGZCNPFKBCDJ-UHFFFAOYSA-N |
Synonym | 2-chloro-6-trifluoromethyl benzoic acid, benzoic acid, 2-chloro-6-trifluoromethyl, 2-chloro-6-trifluoromethyl be, intermediates-zcf02636, 2-trifluoromethyl-6-chlorobenzoic acid, 6-chloro-2-trifluoromethyl benzoic acid, 2-chloro-6-trifluoromethyl benzoic acid, jrd |
PubChem CID | 11127965 |
IUPAC Name | 2-chloro-6-(trifluoromethyl)benzoic acid |
SMILES | C1=CC(=C(C(=C1)Cl)C(=O)O)C(F)(F)F |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH3049003
|
Thermo Scientific Chemicals
H3049003 |
1 g |
Each for $113.68
|
|
|||||
AAH3049006
|
Thermo Scientific Chemicals
H3049006 |
5 g |
Each for $441.45
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
2376-00-3 | |
224.563 | |
OHWGZCNPFKBCDJ-UHFFFAOYSA-N | |
11127965 | |
C1=CC(=C(C(=C1)Cl)C(=O)O)C(F)(F)F |
C8H4ClF3O2 | |
MFCD06208254 | |
2-chloro-6-trifluoromethyl benzoic acid, benzoic acid, 2-chloro-6-trifluoromethyl, 2-chloro-6-trifluoromethyl be, intermediates-zcf02636, 2-trifluoromethyl-6-chlorobenzoic acid, 6-chloro-2-trifluoromethyl benzoic acid, 2-chloro-6-trifluoromethyl benzoic acid, jrd | |
2-chloro-6-(trifluoromethyl)benzoic acid |
Specifications
2376-00-3 | |
C8H4ClF3O2 | |
1 g | |
OHWGZCNPFKBCDJ-UHFFFAOYSA-N | |
2-chloro-6-(trifluoromethyl)benzoic acid | |
11127965 | |
97% |
123°C to 125°C | |
MFCD06208254 | |
2-chloro-6-trifluoromethyl benzoic acid, benzoic acid, 2-chloro-6-trifluoromethyl, 2-chloro-6-trifluoromethyl be, intermediates-zcf02636, 2-trifluoromethyl-6-chlorobenzoic acid, 6-chloro-2-trifluoromethyl benzoic acid, 2-chloro-6-trifluoromethyl benzoic acid, jrd | |
C1=CC(=C(C(=C1)Cl)C(=O)O)C(F)(F)F | |
224.563 | |
224.57 | |
2-Chloro-6-(trifluoromethyl)benzoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures