Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-6-(trifluoromethyl)benzamide, 97%

$89.80 - $358.61

Chemical Identifiers

Chemical Name or Material 2-Chloro-6-(trifluoromethyl)benzamide
CAS 916420-45-6
Molecular Formula C8H5ClF3NO
Formula Weight 223.58
MDL Number MFCD09025362
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3251503

  SDS
alfa aesar™
H3251503
1g Each for $89.80

AAH3251506

  SDS
alfa aesar™
H3251506
5g Each for $358.61
Description & Specifications

Specifications

Formula Weight 223.58
Chemical Name or Material 2-Chloro-6-(trifluoromethyl)benzamide
Assay 97%
Molecular Formula C8H5ClF3NO
MDL Number MFCD09025362
CAS 916420-45-6
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 155° to 158°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.