missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-6-(trifluoromethoxy)aniline, 97%

Manufacturer:  Alfa Aesar™ H3387703

 View more versions of this product

Catalog No. AAH3387703

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-Chloro-6-(trifluoromethoxy)aniline
1261673-70-4
MFCD07779302
2-chloro-6-trifluoromethoxy aniline
C1=CC(=C(C(=C1)Cl)N)OC(F)(F)F
211.568
211.57
97%
1g
C7H5ClF3NO
UN2810
NNDFBODLWFJOSA-UHFFFAOYSA-N
2-chloro-6-(trifluoromethoxy)aniline
86277618
Liquid

Chemical Identifiers

1261673-70-4
211.568
NNDFBODLWFJOSA-UHFFFAOYSA-N
86277618
C1=CC(=C(C(=C1)Cl)N)OC(F)(F)F
C7H5ClF3NO
MFCD07779302
2-chloro-6-trifluoromethoxy aniline
2-chloro-6-(trifluoromethoxy)aniline
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

SDS Documents