Learn More
2-Chloro-6-(trichloromethyl)pyridine, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6641806
Description
2-Chloro-6-(trichloromethyl)pyridine is
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Chloro-6-(trichloromethyl)pyridine is
Solubility
Soluble in 10 mg/ml ethanol.
Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
Specifications
2-Chloro-6-(trichloromethyl)pyridine | |
60°C to 65°C | |
100°C (212°F) | |
MFCD00072500 | |
UN3077 | |
2-chloro-6-trichloromethyl pyridine, nitrapyrin, n-serve, pyridine, 2-chloro-6-trichloromethyl, dowco-163, n-serve nitrogen stabilizer, caswell no. 217, nitrapyrine iso-french, unii-8pce86u01w, nitrapyrin ansi:bsi:iso | |
DCUJJWWUNKIJPH-UHFFFAOYSA-N | |
2-chloro-6-(trichloromethyl)pyridine | |
16004 | |
230.9 | |
Powder |
1929-82-4 | |
White | |
C6H3Cl4N | |
5 g | |
1618997 | |
Soluble in 10mg/ml ethanol. | |
ClC1=CC=CC(=N1)C(Cl)(Cl)Cl | |
230.90 | |
CHEBI:81935 | |
98% |
Chemical Identifiers
1929-82-4 | |
230.90 | |
DCUJJWWUNKIJPH-UHFFFAOYSA-N | |
16004 | |
2-chloro-6-(trichloromethyl)pyridine |
C6H3Cl4N | |
MFCD00072500 | |
2-chloro-6-trichloromethyl pyridine, nitrapyrin, n-serve, pyridine, 2-chloro-6-trichloromethyl, dowco-163, n-serve nitrogen stabilizer, caswell no. 217, nitrapyrine iso-french, unii-8pce86u01w, nitrapyrin ansi:bsi:iso | |
CHEBI:81935 | |
ClC1=CC=CC(=N1)C(Cl)(Cl)Cl |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
DOTInformation : Transport Hazard Class: 9; Packaging Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber : 217-682-2
RTECSNumber : US7525000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only