Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-6-phenoxybenzonitrile, 96%

$256.59 - $256.59

Chemical Identifiers

Chemical Name or Material 2-Chloro-6-phenoxybenzonitrile
CAS 91692-70-5
Molecular Formula C13H8ClNO
Formula Weight 229.67g/mol
MDL Number MFCD00051626
Beilstein 2728571
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0743606

 
alfa aesar™
L07436-06
5g Each for $256.59
Description & Specifications

Specifications

Melting Point 58° to 63°C
Quantity 5g
Chemical Name or Material 2-Chloro-6-phenoxybenzonitrile
CAS 91692-70-5
Assay 96%
Molecular Formula C13H8ClNO
Formula Weight 229.67g/mol
MDL Number MFCD00051626
Beilstein 2728571
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H332
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.