Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-6-nitrotoluene, 98+%

Chemical Identifiers

Chemical Name or Material 2-Chloro-6-nitrotoluene
CAS 83-42-1
Molecular Formula C7H6ClNO2
Formula Weight 171.58
MDL Number MFCD00007205
UN Number UN3457
Beilstein 1239924
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB236750B

  SDS
alfa aesar™
B236750B
1000g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2367530

  SDS
alfa aesar™
B2367530
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 171.58
RTECS Number XS9130000
Boiling Point 238°C
UN Number UN3457
Beilstein 1239924
Flash Point 125°C (257°F)
Quantity 1000g
Chemical Name or Material 2-Chloro-6-nitrotoluene
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CHLORONITROTOLUENES, SOLID
MDL Number MFCD00007205
Melting Point 34° to 38°C
TSCA TSCA
CAS 83-42-1
Health Hazard 3 GHS P Statement
P260-P261-P280-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H331-H373-H302-H312-H315-H319
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Assay ≥98%
Molecular Formula C7H6ClNO2
Health Hazard 1 Danger
Safety and Handling

Danger

GHS P Statement
P260-P261-P280-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H373-H302-H312-H315-H319
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.