missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-6-nitrobenzamide, 98%

$47.35 - $153.12

Chemical Identifiers

CAS 107485-64-3
Molecular Formula C7H5ClN2O3
Molecular Weight (g/mol) 200.578
MDL Number MFCD02093931
InChI Key WPVXBESIHVJEOI-UHFFFAOYSA-N
Synonym 2-chloro-6-nitro-benzamide, benzamide, 2-chloro-6-nitro, benzamide,2-chloro-6-nitro, acmc-20anyb, 6-chloro-2-nitrobenzamide
PubChem CID 86279
IUPAC Name 2-chloro-6-nitrobenzamide
SMILES C1=CC(=C(C(=C1)Cl)C(=O)N)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1694903 View Documents Alfa Aesar™
L1694903
1g Each for $47.35
 
AAL1694906 View Documents Alfa Aesar™
L1694906
5g Each for $153.12
 
Specifications

Chemical Identifiers

107485-64-3
200.578
WPVXBESIHVJEOI-UHFFFAOYSA-N
86279
C1=CC(=C(C(=C1)Cl)C(=O)N)[N+](=O)[O-]
C7H5ClN2O3
MFCD02093931
2-chloro-6-nitro-benzamide, benzamide, 2-chloro-6-nitro, benzamide,2-chloro-6-nitro, acmc-20anyb, 6-chloro-2-nitrobenzamide
2-chloro-6-nitrobenzamide

Specifications

~185°C
107485-64-3
MFCD02093931
2-chloro-6-nitro-benzamide, benzamide, 2-chloro-6-nitro, benzamide,2-chloro-6-nitro, acmc-20anyb, 6-chloro-2-nitrobenzamide
C1=CC(=C(C(=C1)Cl)C(=O)N)[N+](=O)[O-]
200.578
200.58
2-Chloro-6-nitrobenzamide
1g
C7H5ClN2O3
3531056
WPVXBESIHVJEOI-UHFFFAOYSA-N
2-chloro-6-nitrobenzamide
86279
98%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning