Please login to your online account to display your discounted pricing

2-Chloro-6-nitrobenzaldehyde, 98%, Acros Organics

Chemical Identifiers

Chemical Name or Material 2-Chloro-6-nitrobenzaldehyde
Assay Percent Range 98%
Molecular Formula C7H4ClNO3
Formula Weight 185.57
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC147750010

  SDS
thermo fisher scientific
AC147750010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC147750050

  SDS
thermo fisher scientific
AC147750050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 2-Chloro-6-nitrobenzaldehyde
CAS 6361-22-4
Packaging Glass bottle
Quantity 1g
Assay Percent Range 98%
Molecular Formula C7H4ClNO3
Formula Weight 185.57