missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-chloro-6-methylpyrimidine-4-carboxylic acid, 95%, ACROS Organics™

Manufacturer:  Acros Organics AC434300010

 View more versions of this product

Catalog No. AC434300010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-Chloro-6-methylpyrimidine-4-carboxylic acid
Glass bottle
1g
94.0
95%
MFCD00833002
FBXGLOGQWMGIER-UHFFFAOYSA-N
2-chloro-6-methylpyrimidine-4-carboxylic acid
10535213
Crystalline Powder
Authentic
White to Yellow
89581-58-8
100.0
C6H5ClN2O2
2-chloro-6-methyl-pyrimidine-4-carboxylic acid, 2-chloro-6-methyl-4-pyrimidinecarboxylic acid, 2-chloro-4-methylpyrimidine-6-carboxylic acid, 4-pyrimidinecarboxylic acid, 2-chloro-6-methyl
CC1=NC(=NC(=C1)C(=O)O)Cl
172.568
172.57
≥94.0%

Chemical Identifiers

89581-58-8
172.568
FBXGLOGQWMGIER-UHFFFAOYSA-N
10535213
CC1=NC(=NC(=C1)C(=O)O)Cl
C6H5ClN2O2
MFCD00833002
2-chloro-6-methyl-pyrimidine-4-carboxylic acid, 2-chloro-6-methyl-4-pyrimidinecarboxylic acid, 2-chloro-4-methylpyrimidine-6-carboxylic acid, 4-pyrimidinecarboxylic acid, 2-chloro-6-methyl
2-chloro-6-methylpyrimidine-4-carboxylic acid
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS Documents