Please login to your online account to display your discounted pricing

2-chloro-6-methylpyrimidine-4-carboxylic acid, 95%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Chloro-6-methylpyrimidine-4-carboxylic acid
CAS 89581-58-8
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94.0%
Assay Percent Range 95%
Molecular Formula C6H5ClN2O2
Formula Weight 172.57
MDL Number MFCD00833002
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC434300010

  SDS
thermo fisher scientific
AC434300010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC434300050

  SDS
thermo fisher scientific
AC434300050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94.0%
Physical Form Crystalline Powder
Formula Weight 172.57
Quantity 1g
Color White to yellow
CAS Max % 100.0
CAS Min % 94.0
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Assay Percent Range 95%
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Chemical Name or Material 2-Chloro-6-methylpyrimidine-4-carboxylic acid
Molecular Formula C6H5ClN2O2
MDL Number MFCD00833002
CAS 89581-58-8
Packaging Glass bottle
Safety and Handling

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes serious eye irritation.