Please login to your online account to display your discounted pricing

2-Chloro-6-methylpyrimidine-4,5-diamine, ≥95%, Maybridge

Chemical Identifiers

Chemical Name or Material 2-Chloro-6-methylpyrimidine-4, 5-diamine
CAS 63211-98-3
CAS Min % 95.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C5H7ClN4
Formula Weight 158.59
MDL Number MFCD01764685
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

RJC03992CB

  SDS
fisher scientific company
RJC03992CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 158.59
Quantity 250mg
Assay 95%
CAS Min % 95.0
Chemical Name or Material 2-Chloro-6-methylpyrimidine-4, 5-diamine
Packaging Amber Glass Bottle
MDL Number MFCD01764685
CAS 63211-98-3
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if inhaled.
Harmful in contact with skin.
Assay Percent Range 95%
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 3 GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C5H7ClN4