Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-6-methylpyridine-4-carboxylic acid, 98%

$38.49 - $357.11

Chemical Identifiers

Chemical Name or Material 2-Chloro-6-methylpyridine-4-carboxylic acid
CAS 25462-85-5
Molecular Formula C7H6ClNO2
Formula Weight 171.58
MDL Number MFCD00052830
Beilstein 122809
Synonym 2-Chloro-6-methylisonicotinic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1054403

  SDS
alfa aesar™
L1054403
1g Each for $94.81

AAL1054406

  SDS
alfa aesar™
L1054406
5g Each for $357.11

AAL10544MD

  SDS
alfa aesar™
L10544MD
250mg Each for $38.49
Description & Specifications

Specifications

Formula Weight 171.58
Chemical Name or Material 2-Chloro-6-methylpyridine-4-carboxylic acid
Assay 98%
Molecular Formula C7H6ClNO2
CAS 25462-85-5
Beilstein 122809
Synonym 2-Chloro-6-methylisonicotinic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point ≈200°C
MDL Number MFCD00052830
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.