missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-6-methylpyridine-4-carboxylic acid, 98%

Manufacturer:  Alfa Aesar™ L10544MD

 View more versions of this product

Catalog No. AAL10544MD



Specifications

Specifications

2-Chloro-6-methylpyridine-4-carboxylic acid
250mg
C7H6ClNO2
122809
ZGZMEKHQIZSZOH-UHFFFAOYSA-N
2-chloro-6-methylpyridine-4-carboxylic acid
141209
98%
~200°C
25462-85-5
MFCD00052830
2-chloro-6-methylisonicotinic acid, 2-chloro-6-picoline-4-carboxylic acid, 4-pyridinecarboxylic acid, 2-chloro-6-methyl, 2-methyl-6-chloropyridine-4-carboxylic acid, 2-chloro-6-methyl-pyridine-4-carboxylic acid, 2-chloro-6-methyl-4-pyridinecarboxylic acid, 2-chloro-6-methyl-isonicotinic acid, 2-chloro-6-methyl-4-pyridinecarboxylicacid, 2-chloro-6-methylisonicotinicacid, pubchem17665
CC1=NC(=CC(=C1)C(=O)O)Cl
171.58
171.58

Chemical Identifiers

25462-85-5
171.58
ZGZMEKHQIZSZOH-UHFFFAOYSA-N
141209
CC1=NC(=CC(=C1)C(=O)O)Cl
C7H6ClNO2
MFCD00052830
2-chloro-6-methylisonicotinic acid, 2-chloro-6-picoline-4-carboxylic acid, 4-pyridinecarboxylic acid, 2-chloro-6-methyl, 2-methyl-6-chloropyridine-4-carboxylic acid, 2-chloro-6-methyl-pyridine-4-carboxylic acid, 2-chloro-6-methyl-4-pyridinecarboxylic acid, 2-chloro-6-methyl-isonicotinic acid, 2-chloro-6-methyl-4-pyridinecarboxylicacid, 2-chloro-6-methylisonicotinicacid, pubchem17665
2-chloro-6-methylpyridine-4-carboxylic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents