Please login to your online account to display your discounted pricing

2-Chloro-6-methylphenyl isothiocyanate, 95+%

Chemical Identifiers

Chemical Name or Material 2-Chloro-6-methylphenyl isothiocyanate
Assay 95%
Assay Percent Range 95%
Molecular Formula C8H6ClNS
Formula Weight 183.66
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SB01714DA

 
fisher scientific company
SB01714DA
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

SB01714EA

 
fisher scientific company
SB01714EA
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 183.66
Quantity 1g
Assay 95%
Chemical Name or Material 2-Chloro-6-methylphenyl isothiocyanate
Assay Percent Range 95%
Molecular Formula C8H6ClNS
CAS 19241-34-0