Please login to your online account to display your discounted pricing

2-Chloro-6-methylphenyl isothiocyanate, 95+%


Quantity 1g

Manufacturer: fisher scientific company  SB01714DA

 View more versions of this product

Catalog No. SB01714DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 183.66
Quantity 1g
Assay 95%
Chemical Name or Material 2-Chloro-6-methylphenyl isothiocyanate
Assay Percent Range 95%
Molecular Formula C8H6ClNS
CAS 19241-34-0

Specifications

Chemical Name or Material 2-Chloro-6-methylphenyl isothiocyanate
Assay 95%
Assay Percent Range 95%
Molecular Formula C8H6ClNS
Formula Weight 183.66