missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Chloro-6-methylnicotinic acid, 98+%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 270810250
Specifications
2-Chloro-6-methylnicotinic acid | |
Glass bottle | |
157.0°C to 161.0°C | |
30529-70-5 | |
100.0 | |
C7H6ClNO2 | |
2-chloro-6-methyl nicotinic acid, 2-chloro-6-methyl-3-pyridinecarboxylic acid, 2-chloro-6-methyl-nicotinic acid, 2-chloro-6-methylnicotinic acid, 2-chloro-6-methylnicotinicacid, 3-pyridinecarboxylic acid, 2-chloro-6-methyl, 6-methyl-2-chloro nicotinic acid, acmc-209hgo, ksc223m1b, pubchem11019 | |
CC1=NC(=C(C=C1)C(=O)O)Cl | |
171.58 | |
171.58 | |
98+% |
Authentic | |
Undesignated | |
25g | |
98.0 | |
98% min. (HPLC) | |
MFCD00134165 | |
ACQXHCHKMFYDPM-UHFFFAOYSA-N | |
2-chloro-6-methylpyridine-3-carboxylic acid | |
121724 | |
Crystalline Powder |
Chemical Identifiers
30529-70-5 | |
171.58 | |
ACQXHCHKMFYDPM-UHFFFAOYSA-N | |
121724 | |
CC1=NC(=C(C=C1)C(=O)O)Cl |
C7H6ClNO2 | |
MFCD00134165 | |
2-chloro-6-methyl nicotinic acid, 2-chloro-6-methyl-3-pyridinecarboxylic acid, 2-chloro-6-methyl-nicotinic acid, 2-chloro-6-methylnicotinic acid, 2-chloro-6-methylnicotinicacid, 3-pyridinecarboxylic acid, 2-chloro-6-methyl, 6-methyl-2-chloro nicotinic acid, acmc-209hgo, ksc223m1b, pubchem11019 | |
2-chloro-6-methylpyridine-3-carboxylic acid |
Safety and Handling
EINECSNumber : 250-229-7