Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-6-methylnicotinic acid, 97%

$71.27 - $952.27

Chemical Identifiers

Chemical Name or Material 2-Chloro-6-methylnicotinic acid
CAS 30529-70-5
Molecular Formula C7H6ClNO2
Formula Weight 171.58
MDL Number MFCD00134165
Beilstein 473070
Synonym 3-Carboxy-2-chloro-6-methylpyridine; 2-Chloro-6-methylpyridine-3-carboxylic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1385706

  SDS
alfa aesar™
A1385706
5g Each for $71.27

AAA1385714

  SDS
alfa aesar™
A1385714
25g Each for $284.78

AAA1385722

  SDS
alfa aesar™
A1385722
100g Each for $952.27
Description & Specifications

Specifications

Formula Weight 171.58
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g
Assay 97%
Molecular Formula C7H6ClNO2
Melting Point 157° to 162°C
Beilstein 473070
Chemical Name or Material 2-Chloro-6-methylnicotinic acid
MDL Number MFCD00134165
CAS 30529-70-5
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Synonym 3-Carboxy-2-chloro-6-methylpyridine; 2-Chloro-6-methylpyridine-3-carboxylic acid
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.