missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chloro-6-methylaniline, ACROS Organics™

Chemical Identifiers

CAS 87-63-8
Molecular Formula C7H8ClN
Linear Formula ClC6H3(CH3)NH2
Formula Weight 141.6
MDL Number MFCD00007679
Synonym 2-Amino-3-chlorotoluene
Chemical Name or Material 2-Chloro-6-methylaniline
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC109360100 SDS Thermo Fisher Scientific
AC109360100
Glass bottle 10g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

87-63-8
ClC6H3(CH3)NH2
MFCD00007679
2-Chloro-6-methylaniline
C7H8ClN
141.6
2-Amino-3-chlorotoluene

Specifications

1.1520g/mL
87-63-8
C7H8ClN
141.6
2-Amino-3-chlorotoluene
10g
Glass bottle
ClC6H3(CH3)NH2
MFCD00007679
2-Chloro-6-methylaniline

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning