missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-chloro-6-methyl-4-(trifluoromethyl)pyridine, 97%, Maybridge™

Manufacturer:  Fisher Scientific Company CD10452EA

 View more versions of this product

Catalog No. CD10452EA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

2-Chloro-6-methyl-4-(trifluoromethyl)pyridine
10g
96.5
97%
MFCD00052900
SXLBWNSGCIEART-UHFFFAOYSA-N
2-chloro-6-methyl-4-(trifluoromethyl)pyridine
2736626
97%
Amber glass bottle
22123-14-4
100.0
C7H5ClF3N
2-chlor-6-methyl-4-trifluormethyl pyridine, 2-chloranyl-6-methyl-4-trifluoromethyl pyridine, 2-chloro-4-trifluoromethyl-6-methylpyridine, 2-chloro-6-methyl-4-trifluoromethyl pyridine, 2-chloro-6-methyl-4-trifluoromethyl-pyridine, 6-chloro-4-trifluoromethyl-2-picoline, pubchem9219, pyridine, 2-chloro-6-methyl-4-trifluoromethyl
CC1=NC(Cl)=CC(=C1)C(F)(F)F
195.57
195.57

Chemical Identifiers

22123-14-4
195.57
SXLBWNSGCIEART-UHFFFAOYSA-N
2736626
CC1=NC(Cl)=CC(=C1)C(F)(F)F
C7H5ClF3N
MFCD00052900
2-chlor-6-methyl-4-trifluormethyl pyridine, 2-chloranyl-6-methyl-4-trifluoromethyl pyridine, 2-chloro-4-trifluoromethyl-6-methylpyridine, 2-chloro-6-methyl-4-trifluoromethyl pyridine, 2-chloro-6-methyl-4-trifluoromethyl-pyridine, 6-chloro-4-trifluoromethyl-2-picoline, pubchem9219, pyridine, 2-chloro-6-methyl-4-trifluoromethyl
2-chloro-6-methyl-4-(trifluoromethyl)pyridine
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS Documents
Product Certifications