Please login to your online account to display your discounted pricing

2-chloro-6-methyl-4-(trifluoromethyl)pyridine, 97%, Maybridge


Quantity 5g

Manufacturer: fisher scientific company  CD10452DE

 View more versions of this product

Catalog No. CD10452DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 195.57
Quantity 5g
Assay 97%
CAS Max % 100.0
CAS 22123-14-4
Molecular Formula C7H5ClF3N
Chemical Name or Material 2-Chloro-6-methyl-4-(trifluoromethyl)pyridine
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00052900
CAS Min % 96.5
Assay Percent Range 97%
Health Hazard 1 Warning

Specifications

Chemical Name or Material 2-Chloro-6-methyl-4-(trifluoromethyl)pyridine
CAS 22123-14-4
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C7H5ClF3N
Formula Weight 195.57
MDL Number MFCD00052900
Accessories

Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.