missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Alfa Aesar™ 2-Chloro-6-methyl-4-nitroaniline, 98%
$43.19 - $149.78
Chemical Identifiers
CAS | 69951-02-6 |
---|---|
Molecular Formula | C7H7ClN2O2 |
Molecular Weight (g/mol) | 186.595 |
MDL Number | MFCD01313703 |
InChI Key | HVPVUAIKGOKEEE-UHFFFAOYSA-N |
Synonym | 2-chloro-4-nitro-6-methylaniline, 2-chloro-6-methyl-4-nitro-phenylamine, 2-chloro-6-methyl-4-nitrophenylamine, 6-chloro-2-methyl-4-nitroaniline, 6-chloro-2-methyl-4-nitrophenylamine, acmc-1b462, benzenamine, 2-chloro-6-methyl-4-nitro, benzenamine,2-chloro-6-methyl-4-nitro, chloromethylnitroaniline, pubchem4373 |
PubChem CID | 2733460 |
IUPAC Name | 2-chloro-6-methyl-4-nitroaniline |
SMILES | CC1=CC(=CC(=C1N)Cl)[N+](=O)[O-] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2181403
|
Alfa Aesar™
B2181403 |
1g |
Each for $43.19
|
|
AAB2181406
|
Alfa Aesar™
B2181406 |
5g |
Each for $149.78
|
|
Chemical Identifiers
69951-02-6 | |
186.595 | |
HVPVUAIKGOKEEE-UHFFFAOYSA-N | |
2733460 | |
CC1=CC(=CC(=C1N)Cl)[N+](=O)[O-] |
C7H7ClN2O2 | |
MFCD01313703 | |
2-chloro-4-nitro-6-methylaniline, 2-chloro-6-methyl-4-nitro-phenylamine, 2-chloro-6-methyl-4-nitrophenylamine, 6-chloro-2-methyl-4-nitroaniline, 6-chloro-2-methyl-4-nitrophenylamine, acmc-1b462, benzenamine, 2-chloro-6-methyl-4-nitro, benzenamine,2-chloro-6-methyl-4-nitro, chloromethylnitroaniline, pubchem4373 | |
2-chloro-6-methyl-4-nitroaniline |
Specifications
174°C to 176°C | |
69951-02-6 | |
MFCD01313703 | |
HVPVUAIKGOKEEE-UHFFFAOYSA-N | |
2-chloro-6-methyl-4-nitroaniline | |
2733460 | |
98% |
1g | |
C7H7ClN2O2 | |
2-chloro-4-nitro-6-methylaniline, 2-chloro-6-methyl-4-nitro-phenylamine, 2-chloro-6-methyl-4-nitrophenylamine, 6-chloro-2-methyl-4-nitroaniline, 6-chloro-2-methyl-4-nitrophenylamine, acmc-1b462, benzenamine, 2-chloro-6-methyl-4-nitro, benzenamine,2-chloro-6-methyl-4-nitro, chloromethylnitroaniline, pubchem4373 | |
CC1=CC(=CC(=C1N)Cl)[N+](=O)[O-] | |
186.595 | |
186.6 | |
2-Chloro-6-methyl-4-nitroaniline |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures