missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chloro-6-methyl-3-(trifluoromethyl)pyridine, 97%, ACROS Organics™

Chemical Identifiers

CAS 1099597-74-6
Molecular Formula C7H5ClF3N
Molecular Weight (g/mol) 195.57
InChI Key DIFCQOVMVHCCHY-UHFFFAOYSA-N
Synonym 2-chloro-3-trifluoromethyl-6-methylpyridine, 2-chloro-6-methyl-3-trifluoromethyl pyridine, pyridine, 2-?chloro-?6-?methyl-?3-? trifluoromethyl ?, pyridine, 2-chloro-6-methyl-3-trifluoromethyl
PubChem CID 37863457
IUPAC Name 2-chloro-6-methyl-3-(trifluoromethyl)pyridine
SMILES CC1=NC(=C(C=C1)C(F)(F)F)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC452440010
SDS View Documents
Thermo Fisher Scientific
AC452440010
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC452440050
SDS View Documents
Thermo Fisher Scientific
AC452440050
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

1099597-74-6
195.57
2-chloro-3-trifluoromethyl-6-methylpyridine, 2-chloro-6-methyl-3-trifluoromethyl pyridine, pyridine, 2-?chloro-?6-?methyl-?3-? trifluoromethyl ?, pyridine, 2-chloro-6-methyl-3-trifluoromethyl
2-chloro-6-methyl-3-(trifluoromethyl)pyridine
C7H5ClF3N
DIFCQOVMVHCCHY-UHFFFAOYSA-N
37863457
CC1=NC(=C(C=C1)C(F)(F)F)Cl

Specifications

Glass Bottle
30°C to 32°C
1099597-74-6
C7H5ClF3N
DIFCQOVMVHCCHY-UHFFFAOYSA-N
2-chloro-6-methyl-3-(trifluoromethyl)pyridine
37863457
97%
84°C to 86°C (24.0 Torr)
1g
96% min. (GC)
2-chloro-3-trifluoromethyl-6-methylpyridine, 2-chloro-6-methyl-3-trifluoromethyl pyridine, pyridine, 2-?chloro-?6-?methyl-?3-? trifluoromethyl ?, pyridine, 2-chloro-6-methyl-3-trifluoromethyl
CC1=NC(=C(C=C1)C(F)(F)F)Cl
195.57
195.57
2-Chloro-6-methyl-3-(trifluoromethyl)pyridine
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS