missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Chloro-6-methyl-3-(trifluoromethyl)pyridine, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC452440050
Specifications
2-Chloro-6-methyl-3-(trifluoromethyl)pyridine | |
30°C to 32°C | |
1099597-74-6 | |
C7H5ClF3N | |
DIFCQOVMVHCCHY-UHFFFAOYSA-N | |
2-chloro-6-methyl-3-(trifluoromethyl)pyridine | |
37863457 | |
97% |
84°C to 86°C (24.0 Torr) | |
5g | |
96% min. (GC) | |
2-chloro-3-trifluoromethyl-6-methylpyridine, 2-chloro-6-methyl-3-trifluoromethyl pyridine, pyridine, 2-?chloro-?6-?methyl-?3-? trifluoromethyl ?, pyridine, 2-chloro-6-methyl-3-trifluoromethyl | |
CC1=NC(=C(C=C1)C(F)(F)F)Cl | |
195.57 | |
195.57 |
Chemical Identifiers
1099597-74-6 | |
195.57 | |
2-chloro-3-trifluoromethyl-6-methylpyridine, 2-chloro-6-methyl-3-trifluoromethyl pyridine, pyridine, 2-?chloro-?6-?methyl-?3-? trifluoromethyl ?, pyridine, 2-chloro-6-methyl-3-trifluoromethyl | |
2-chloro-6-methyl-3-(trifluoromethyl)pyridine |
C7H5ClF3N | |
DIFCQOVMVHCCHY-UHFFFAOYSA-N | |
37863457 | |
CC1=NC(=C(C=C1)C(F)(F)F)Cl |
Safety and Handling
GHS Signal Word: Warning