Please login to your online account to display your discounted pricing

2-Chloro-6-methyl-3-pyridinecarbonitrile, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Chloro-6-methyl-3-pyridinecarbonitrile
Name Note 99%
CAS 28900-10-9
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7H5ClN2
Formula Weight 152.58
MDL Number MFCD00082767
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC295610010

  SDS
thermo fisher scientific
AC295610010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Crystalline Powder
Formula Weight 152.58
Quantity 1g
Color Beige
TSCA TSCA
Melting Point 114° to 117°C
CAS Max % 100.0
Chemical Name or Material 2-Chloro-6-methyl-3-pyridinecarbonitrile
MDL Number MFCD00082767
CAS Min % 98.5
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.
Harmful in contact with skin.
Infrared Spectrum Authentic
Packaging Glass bottle
Name Note 99%
Assay Percent Range 99%
Molecular Formula C7H5ClN2
CAS 28900-10-9
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.
Harmful in contact with skin.