Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-6-methoxyquinoline-3-carboxaldehyde, 99%

$93.27 - $320.18

Chemical Identifiers

Chemical Name or Material 2-Chloro-6-methoxyquinoline-3-carboxaldehyde
CAS 73568-29-3
Molecular Formula C11H8ClNO2
Formula Weight 221.64
MDL Number MFCD00160585
Synonym 2-Chloro-3-formyl-6-methoxyquinoline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2697306

 
alfa aesar™
H2697306
5g Each for $93.27

AAH2697314

 
alfa aesar™
H2697314
25g Each for $320.18
Description & Specifications

Specifications

Formula Weight 221.64
Chemical Name or Material 2-Chloro-6-methoxyquinoline-3-carboxaldehyde
Assay 99%
Molecular Formula C11H8ClNO2
MDL Number MFCD00160585
CAS 73568-29-3
Synonym 2-Chloro-3-formyl-6-methoxyquinoline
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point −9°C (16°F)
Melting Point 146° to 151°C
Quantity 5g

Applications
Mainly used as solvents. They are used in intermediate steps for the production of paints, plastics, synthetic resins, and dyes. They are also used in the manufacture of perfumes, solvents, and flavorings. As raw material in organic synthesis.

Solubility
Insoluble in water.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.