Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-6-methoxypyridine-4-carboxylic acid, 96%

$86.19 - $311.91

Chemical Identifiers

Chemical Name or Material 2-Chloro-6-methoxypyridine-4-carboxylic acid
CAS 15855-06-8
Molecular Formula C7H6ClNO3
Formula Weight 187.58
MDL Number MFCD00173928
Synonym 2-Chloro-6-methoxyisonicotinic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5016803

  SDS
alfa aesar™
H5016803
1g Each for $86.19

AAH5016806

  SDS
alfa aesar™
H5016806
5g Each for $311.91
Description & Specifications

Specifications

Formula Weight 187.58
Chemical Name or Material 2-Chloro-6-methoxypyridine-4-carboxylic acid
Assay 96%
MDL Number MFCD00173928
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
Synonym 2-Chloro-6-methoxyisonicotinic acid
Boiling Point 209° to 213°C
Molecular Formula C7H6ClNO3
CAS 15855-06-8
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.