Please login to your online account to display your discounted pricing

2-Chloro-6-fluorotoluene 97%, ACROS Organics™


Quantity 250g

Manufacturer: thermo fisher scientific  161502500

 View more versions of this product

Catalog No. AC161502500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥96.0%
Formula Weight 144.58
Quantity 250g
Color Clear colorless
EINECS Number 207-141-9
TSCA TSCA
CAS Min % 96.0
CAS Max % 100.0
Refractive Index 1.5020 to 1.5040 (20°C, 589nm)
Chemical Name or Material 2-Chloro-6-fluorotoluene
Name Note 97%
CAS 443-83-4
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Assay Percent Range 97%
Molecular Formula C7 H6 Cl F
MDL Number MFCD00000570
Synonym 1-Chloro-3-fluoro-2-methylbenzene
Packaging Glass bottle

Specifications

Chemical Name or Material 2-Chloro-6-fluorotoluene
Name Note 97%
CAS 443-83-4
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C7 H6 Cl F
Formula Weight 144.58
MDL Number MFCD00000570
Synonym 1-Chloro-3-fluoro-2-methylbenzene
Accessories

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Flammable liquid and vapor.

SDS & Safety Documents