missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chloro-6-fluorotoluene 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 161500500

 View more versions of this product

Catalog No. AC161500500

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

2-Chloro-6-fluorotoluene
Clear colorless
97%
96.0
100.0
Liquid
97%
144.58
1-Chloro-3-fluoro-2-methylbenzene
Authentic
50g
443-83-4
Glass bottle
1.5020 to 1.5040 (20°C, 589nm)
≥96.0%
C7 H6 Cl F
MFCD00000570

Chemical Identifiers

443-83-4
100.0
C7 H6 Cl F
MFCD00000570
2-Chloro-6-fluorotoluene, 97%
96.0
97%
144.58
1-Chloro-3-fluoro-2-methylbenzene
≥96.0%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 207-141-9

TSCA : TSCA