missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chloro-6-fluorotoluene 97%, ACROS Organics™

Chemical Identifiers

CAS 443-83-4
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C7 H6 Cl F
Formula Weight 144.58
MDL Number MFCD00000570
Synonym 1-Chloro-3-fluoro-2-methylbenzene
Chemical Name or Material 2-Chloro-6-fluorotoluene, 97%
Assay ≥96.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC161500500 SDS Thermo Fisher Scientific
161500500
50g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC161502500 SDS Thermo Fisher Scientific
161502500
250g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

443-83-4
100.0
C7 H6 Cl F
MFCD00000570
2-Chloro-6-fluorotoluene, 97%
96.0
97%
144.58
1-Chloro-3-fluoro-2-methylbenzene
≥96.0%

Specifications

Liquid
50g
100.0
Authentic
C7 H6 Cl F
443-83-4
Glass bottle
2-Chloro-6-fluorotoluene, 97%
144.58
Clear colorless
96.0
MFCD00000570
97%
1.5020 to 1.5040 (20°C, 589nm)
1-Chloro-3-fluoro-2-methylbenzene
≥96.0%

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 207-141-9

TSCA : TSCA