missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chloro-6-fluorophenylacetic Acid, TCI America™

$48.89 - $160.42

Chemical Identifiers

CAS 37777-76-7
Molecular Formula C8H6ClFO2
Molecular Weight (g/mol) 188.582
MDL Number MFCD00004319
InChI Key GUAIAAXDEJZRBP-UHFFFAOYSA-N
Synonym 2-chloro-6-fluorophenylacetic acid, 2-2-chloro-6-fluorophenyl acetic acid, 2-chloro-6-fluorophenyl acetic acid, 2-chloro-6-fluorophenylaceticacid, 2-chloro-6-fluorophenyl acetic acid, benzeneacetic acid, 2-chloro-6-fluoro, 2-chloro-6-fluoro-phenyl-acetic acid, pubchem4254, acmc-1aewm, rarechem al bo 0114
PubChem CID 123464
IUPAC Name 2-(2-chloro-6-fluorophenyl)acetic acid
SMILES C1=CC(=C(C(=C1)Cl)CC(=O)O)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C11265G View Documents TCI America
C11265G
5g Each for $48.89
 
C112625G View Documents TCI America
C112625G
25g Each for $160.42
 
Specifications

Chemical Identifiers

37777-76-7
188.582
GUAIAAXDEJZRBP-UHFFFAOYSA-N
123464
C1=CC(=C(C(=C1)Cl)CC(=O)O)F
C8H6ClFO2
MFCD00004319
2-chloro-6-fluorophenylacetic acid, 2-2-chloro-6-fluorophenyl acetic acid, 2-chloro-6-fluorophenyl acetic acid, 2-chloro-6-fluorophenylaceticacid, 2-chloro-6-fluorophenyl acetic acid, benzeneacetic acid, 2-chloro-6-fluoro, 2-chloro-6-fluoro-phenyl-acetic acid, pubchem4254, acmc-1aewm, rarechem al bo 0114
2-(2-chloro-6-fluorophenyl)acetic acid

Specifications

White-Yellow
5g
C8H6ClFO2
2-chloro-6-fluorophenylacetic acid, 2-2-chloro-6-fluorophenyl acetic acid, 2-chloro-6-fluorophenyl acetic acid, 2-chloro-6-fluorophenylaceticacid, 2-chloro-6-fluorophenyl acetic acid, benzeneacetic acid, 2-chloro-6-fluoro, 2-chloro-6-fluoro-phenyl-acetic acid, pubchem4254, acmc-1aewm, rarechem al bo 0114
C1=CC(=C(C(=C1)Cl)CC(=O)O)F
188.582
188.58
≥98.0% (T)
123°C
37777-76-7
MFCD00004319
GUAIAAXDEJZRBP-UHFFFAOYSA-N
2-(2-chloro-6-fluorophenyl)acetic acid
123464
Crystal-Powder at 20°C
2-Chloro-6-fluorophenylacetic Acid
Safety and Handling

Safety and Handling

TSCA : No