Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-6-fluorophenol, 97%

Chemical Identifiers

Chemical Name or Material 2-Chloro-6-fluorophenol
CAS 2040-90-6
Molecular Formula C6H4ClFO
Formula Weight 146.55
MDL Number MFCD01631574
UN Number UN3261
Beilstein 1936441
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2330303

  SDS
alfa aesar™
B2330303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2330306

  SDS
alfa aesar™
B2330306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 146.55
Chemical Name or Material 2-Chloro-6-fluorophenol
Assay 97%
Molecular Formula C6H4ClFO
MDL Number MFCD01631574
CAS 2040-90-6
UN Number UN3261
Beilstein 1936441
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H340-H361-H314-H318-H302-H317
May cause genetic defects.
Suspected of damaging fertility or the unborn child.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement
P273-P201-P309-P310
Avoid release to the environment.
Obtain special instructions before use.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Odor Phenol-like
Melting Point 61° to 65°C
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P273-P201-P309-P310
Avoid release to the environment.
Obtain special instructions before use.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H340-H361-H314-H318-H302-H317
May cause genetic defects.
Suspected of damaging fertility or the unborn child.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.