missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-6-fluorobenzylideneacetone, 98%

Manufacturer:  Alfa Aesar™ A1508314

 View more versions of this product

Catalog No. AAA1508314



Specifications

Specifications

2-Chloro-6-fluorobenzylideneacetone
25g
C10H8ClFO
2-chloro-6-fluorobenzylideneacetone, 3e-4-2-chloro-6-fluorophenyl but-3-en-2-one, 4-2-chloro-6-fluorophenyl but-3-en-2-one, 1-2-chloro-6-fluorophenyl but-1-en-3-one, e-4-2-chloro-6-fluoro-phenyl but-3-en-2-one, labotest-bb lt00455271, 1-2-chloro-6-fluorophenyl-1-buten-2-one, e-4-2-chloro-6-fluorophenyl-3-buten-2-one, e-4-2-chloro-6-fluorophenyl but-3-en-2-one, 1-2-chloro-6-fluorophenyl but-1-en-3-one
CC(=O)C=CC1=C(C=CC=C1Cl)F
198.621
198.63
115°C (0.8mmHg)
175136-82-0
MFCD00051608
VZRBDBXFAKDNDJ-AATRIKPKSA-N
(E)-4-(2-chloro-6-fluorophenyl)but-3-en-2-one
5373895
98%

Chemical Identifiers

175136-82-0
198.621
VZRBDBXFAKDNDJ-AATRIKPKSA-N
5373895
CC(=O)C=CC1=C(C=CC=C1Cl)F
C10H8ClFO
MFCD00051608
2-chloro-6-fluorobenzylideneacetone, 3e-4-2-chloro-6-fluorophenyl but-3-en-2-one, 4-2-chloro-6-fluorophenyl but-3-en-2-one, 1-2-chloro-6-fluorophenyl but-1-en-3-one, e-4-2-chloro-6-fluoro-phenyl but-3-en-2-one, labotest-bb lt00455271, 1-2-chloro-6-fluorophenyl-1-buten-2-one, e-4-2-chloro-6-fluorophenyl-3-buten-2-one, e-4-2-chloro-6-fluorophenyl but-3-en-2-one, 1-2-chloro-6-fluorophenyl but-1-en-3-one
(E)-4-(2-chloro-6-fluorophenyl)but-3-en-2-one
Documents