Please login to your online account to display your discounted pricing

2-chloro-6-fluorobenzylamine, 97%, ACROS Organics™

$94.28 - $94.28

Chemical Identifiers

Chemical Name or Material 2-Chloro-6-fluorobenzylamine
CAS 15205-15-9
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C7H7ClFN
Linear Formula ClC6H3(F)CH2NH2
Formula Weight 159.59
MDL Number MFCD00042458
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC350631000

  SDS
thermo fisher scientific
AC350631000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC350630250

  SDS
thermo fisher scientific
35063-0250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC350630050

  SDS
thermo fisher scientific
35063-0050
Glass bottle 5g Each for $94.28
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Liquid
Formula Weight 159.59
Quantity 100g
Color Clear colorless to light yellow
Density 1.2400g/mL
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 2-Chloro-6-fluorobenzylamine
Infrared Spectrum Authentic
Assay Percent Range 97%
Molecular Formula C7H7ClFN
CAS 15205-15-9
Health Hazard 1 GHS Signal Word: Danger
Packaging Glass bottle
Linear Formula ClC6H3(F)CH2NH2
MDL Number MFCD00042458
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Refractive Index 1.5340 to 1.5360 (20°C, 589nm)
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.