Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-6-fluorobenzyl chloride, 98%

Chemical Identifiers

Chemical Name or Material 2-Chloro-6-fluorobenzyl chloride
CAS 55117-15-2
Molecular Formula C7H5Cl2F
Formula Weight 179.02
MDL Number MFCD00000896
UN Number UN3265
Beilstein 386840
Synonym alpha,2-Dichloro-6-fluorotoluene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1175814

  SDS
alfa aesar™
A1175814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1175822

  SDS
alfa aesar™
A1175822
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 179.02
Health Hazard 1 Danger
TSCA TSCA
Beilstein 386840
Synonym alpha,2-Dichloro-6-fluorotoluene
CAS 55117-15-2
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
Quantity 25g
MDL Number MFCD00000896
Molecular Formula C7H5Cl2F
Chemical Name or Material 2-Chloro-6-fluorobenzyl chloride
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Health Hazard 2 GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Assay 98%
Boiling Point 208° to 210°C
UN Number UN3265
Flash Point 93°C (199°F)
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.