Please login to your online account to display your discounted pricing

2-chloro-6-fluorobenzyl bromide, 96%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-chloro-6-fluorobenzyl bromide
CAS 68220-26-8
CAS Min % 95.0
CAS Max % 100.0
Assay ≥95.0%
Assay Percent Range 96%
Molecular Formula C7H5BrClF
Linear Formula Cl(F)C6H3CH2Br
Formula Weight 223.47
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC373760010

 
thermo fisher scientific
AC373760010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC373760050

 
thermo fisher scientific
AC373760050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Assay ≥95.0%
Formula Weight 223.47
Quantity 1g
Color Colorless to yellow
Density 1.629g/mL
CAS Min % 95.0
CAS Max % 100.0
Linear Formula Cl(F)C6H3CH2Br
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 68220-26-8
Molecular Formula C7H5BrClF
Assay Percent Range 96%
Chemical Name or Material 2-chloro-6-fluorobenzyl bromide
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.