Please login to your online account to display your discounted pricing

2-Chloro-6-fluorobenzyl alcohol, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Chloro-6-fluorobenzyl alcohol
Name Note 97%
CAS 56456-50-9
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C7H6ClFO
Linear Formula ClC6H3(F)CH2OH
Formula Weight 160.57
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC182090250

  SDS
thermo fisher scientific
AC182090250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Crystalline Solid
Formula Weight 160.57
Quantity 25g
Color White
EINECS Number 260-192-9
TSCA TSCA
Melting Point 41° to 44°C
CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
CAS 56456-50-9
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Linear Formula ClC6H3(F)CH2OH
MDL Number MFCD00004608
Chemical Name or Material 2-Chloro-6-fluorobenzyl alcohol
Name Note 97%
Assay Percent Range 97%
Molecular Formula C7H6ClFO
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.